Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039045.1 | NP_001034134.1 | 2 | 1521 | ||
NM_026078.2 | NP_080354.1 | 2 | 1534 | |||
GenBank mRNA | AK014096.1 | - | 2 | 1496 | ||
AK155971.1 | - | 1 | 2444 | |||
AK161117.1 | - | 2 | 1490 |