Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026080.2 | NP_080356.1 | 4 | 528 | ||
GenBank mRNA | AK003117.1 | - | 4 | 512 | ||
AK007338.1 | - | 4 | 512 | |||
AK010590.1 | - | 4 | 509 | |||
AK012461.1 | - | 4 | 517 | |||
AK014099.1 | - | 4 | 515 | |||
AK215321.1 | - | 1 | 155 | |||
BC029262.1 | - | 4 | 502 | 4911699 |