Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctex1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163767.1 | NP_001157239.1 | 5 | 572 | ||
NM_001163768.1 | NP_001157240.1 | 4 | 479 | |||
NM_026100.3 | NP_080376.1 | 5 | 604 | |||
GenBank mRNA | AK006804.1 | - | 5 | 573 | ||
AK015315.1 | - | 5 | 607 | |||
BB634690.1 | - | 4 | 491 | |||
BC048635.1 | - | 5 | 544 | |||
BC099549.1 | - | 5 | 578 | 30919464 | ||
BC132398.1 | - | 5 | 522 | 40130675 | ||
BC132400.1 | - | 5 | 522 | 40130677 | ||
BC145187.1 | - | 5 | 522 | 9053738 |