Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026106.4 | NP_080382.2 | 3 | 1702 | ||
GenBank mRNA | AK031057.1 | - | 3 | 1636 | ||
AK075713.1 | - | 3 | 1702 | |||
AK090113.1 | - | 3 | 1684 | |||
AK144749.1 | - | 3 | 1696 | |||
AK146573.1 | - | 3 | 1665 | |||
BC013461.1 | - | 3 | 1271 | 4222118 |