Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000093.4 | NP_000084.3 | 66 | 6461 | ||
NM_001278074.1 | NP_001265003.1 | 66 | 6461 | |||
GenBank mRNA | AB209864.1 | - | 65 | 5940 | ||
AB371583.1 | - | 66 | 6461 | |||
AK127662.1 | - | 2 | 3773 | |||
BC008760.2 | - | 19 | 2337 | |||
CB152831.1 | - | 1 | 170 |