Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026119.3 | NP_080395.1 | 1, 2 | 145 | ||
GenBank mRNA | AK010270.1 | - | 1, 2 | 147 | ||
AK010686.1 | - | 1, 2 | 147 | |||
AK078288.1 | - | 1, 2 | 157 | |||
AK145636.1 | - | 1, 2 | 170 | |||
BC024950.1 | - | 1, 2 | 145 | 3992287 |