Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026122.4 | NP_080398.1 | 5 | 362 | ||
NM_175074.2 | NP_778249.1 | 5 | 362 | |||
GenBank mRNA | AK002970.1 | - | 5 | 281 | ||
AK003272.1 | - | 5 | 310 | |||
AK153223.1 | - | 5 | 322 | |||
BC005693.1 | - | 5 | 298 | 3597594 |