Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833420G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113550.1 | NP_001107022.1 | 2 | 582 | ||
NM_026127.4 | NP_080403.4 | 3 | 645 | |||
GenBank mRNA | AK008565.1 | - | 2 | 583 | ||
AK008588.1 | - | 2 | 581 | |||
AK014735.1 | - | 2 | 505 | |||
AK146515.1 | - | 2 | 573 | |||
AK149119.1 | - | 3 | 1435 | |||
AK160863.1 | - | 2 | 566 | |||
BC021327.1 | - | 3 | 625 | 5066171 | ||
BC028328.1 | - | 3 | 1414 | |||
BC030738.1 | - | 3 | 613 |