Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhdds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026144.4 | NP_080420.2 | 3 | 315 | ||
GenBank mRNA | AK004993.1 | - | 3 | 377 | ||
AK014317.1 | - | 3 | 377 | |||
AK034277.1 | - | 3 | 377 | |||
AK036929.1 | - | 3 | 393 | |||
AK049991.1 | - | 3 | 316 | |||
AK135087.1 | - | 3 | 377 | |||
AK149138.1 | - | 3 | 347 | |||
AK171207.1 | - | 3 | 389 | |||
BC004011.1 | - | 3 | 363 | 3493585 |