Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159941.1 | NP_001153413.1 | 2, 3 | 302 | ||
NM_026145.4 | NP_080421.2 | 2, 3 | 302 | |||
GenBank mRNA | AK028657.1 | - | 2, 3 | 269 | ||
AK041516.1 | - | 2, 3 | 269 | |||
AK145295.1 | - | 2, 3 | 281 | |||
AK147714.1 | - | 2, 3 | 247 | |||
AK161199.1 | - | 2, 3 | 259 | |||
AK163351.1 | - | 2, 3 | 256 | |||
AK166505.1 | - | 2, 3 | 281 | |||
AK193695.1 | - | 115 | ||||
BB855690.1 | - | 2, 3 | 302 | |||
BC006935.1 | - | 2, 3 | 283 | 3601917 | ||
BQ571808.1 | - | 2, 3 | 253 |