Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026154.1 | NP_080430.1 | 3 | 344 | ||
GenBank mRNA | AK002865.1 | - | 3 | 344 | ||
AK007643.1 | - | 3 | 336 | |||
AK078463.1 | - | 3 | 335 | |||
AK150774.1 | - | 3 | 336 | |||
AK171028.1 | - | 3 | 338 | |||
BC016219.1 | - | 3 | 330 | 4485332 |