Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000097.5 | NP_000088.3 | 5, 6 | 1283 | ||
GenBank mRNA | AK223481.1 | - | 5, 6 | 1213 | ||
AK290140.1 | - | 5, 6 | 1283 | |||
BC017210.2 | - | 5, 6 | 1234 | 3956896 | ||
BC023551.2 | - | 5, 6 | 1269 | 3161428 | ||
BC023554.2 | - | 5, 6 | 1230 | 3607724 | ||
D16611.1 | - | 5, 6 | 969 | |||
Z28409.1 | - | 5, 6 | 876 |