Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1lc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026160.4 | NP_080436.1 | 4 | 482 | ||
GenBank mRNA | AF255953.1 | - | 4 | 500 | ||
AK002795.1 | - | 4 | 485 | |||
AK003106.1 | - | 4 | 483 | |||
AK003205.1 | - | 4 | 483 | |||
AK003558.1 | - | 4 | 485 | |||
AK012604.1 | - | 4 | 486 | |||
AK132329.1 | - | 490 | ||||
AK151614.1 | - | 4 | 481 | |||
AK172322.1 | - | 1 | 41 | |||
AK199755.1 | - | 2 | 255 | |||
BC068180.1 | - | 4 | 485 | 30634082 | ||
BC086774.1 | - | 3 | 2909 | |||
BC110308.1 | - | 4 | 473 | 5319360 |