Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026161.3 | NP_080437.2 | 2 | 984 | ||
GenBank mRNA | AK002948.1 | - | 2 | 911 | ||
AK004340.1 | - | 1 | 533 | |||
AK029777.1 | - | 1 | 159 | |||
BC027315.1 | - | 2 | 832 | |||
BC087832.1 | - | 1 | 4296 | |||
DQ002397.1 | - | 1 | 761 |