Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000108.4 | NP_000099.2 | 7 | 668 | ||
NM_001289750.1 | NP_001276679.1 | 5 | 519 | |||
NM_001289751.1 | NP_001276680.1 | 6 | 599 | |||
GenBank mRNA | AB209703.1 | - | 7 | 578 | ||
AK225289.1 | - | 7 | 592 | |||
AK225301.1 | - | 7 | 604 | |||
AK294146.1 | - | 8 | 605 | |||
AK295080.1 | - | 5 | 462 | |||
AK297508.1 | - | 6 | 533 | |||
AK310220.1 | - | 7 | 684 | |||
AK312346.1 | - | 7 | 615 | |||
BC018648.2 | - | 7 | 577 | 3029752 | ||
BC018696.2 | - | 7 | 577 | 3029752 | ||
BI562331.1 | - | 7 | 643 | |||
J03490.1 | - | 7 | 600 | |||
J03620.1 | - | 7 | 574 |