Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026186.4 | NP_080462.2 | 2 | 254 | ||
GenBank mRNA | AK004983.2 | - | 2 | 198 | ||
AK017849.1 | - | 2 | 199 | |||
AK030628.1 | - | 2 | 183 | |||
AK035158.1 | - | 2 | 183 | |||
BC021478.1 | - | 2 | 168 | |||
BC079553.1 | - | 2 | 176 | 6856335 | ||
BY307671.1 | - | 2 | 254 |