Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110008F13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026124.3 | NP_080400.1 | 2, 3 | 407 | ||
GenBank mRNA | AK002452.1 | - | 2, 3 | 365 | ||
AK003568.1 | - | 2, 3 | 402 | |||
AK179243.1 | - | 2, 3 | 128 | |||
AK186989.1 | - | 2, 3 | 128 | |||
BC019445.1 | - | 2, 3 | 345 | 5125733 |