Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armcx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166397.1 | NP_001159869.1 | 6 | 1280 | ||
NM_001166398.1 | NP_001159870.1 | 6 | 1223 | |||
NM_026139.4 | NP_080415.3 | 5 | 1387 | |||
GenBank mRNA | AK014329.1 | - | 6 | 1056 | ||
AK034185.1 | - | 6 | 1056 | |||
AK034621.1 | - | 6 | 1054 | |||
AK043440.1 | - | 6 | 1056 | |||
AK044926.1 | - | 6 | 1054 | |||
AK053354.1 | - | 5 | 1161 | |||
AK081299.1 | - | 6 | 999 | |||
AK172960.1 | - | 6 | 1280 | |||
BC049088.1 | - | 6 | 1035 | |||
BC052383.1 | - | 6 | 1029 | |||
BC054839.1 | - | 3 | 619 | 6490133 |