Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ears2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014324.2 | - | 1153 | |||
AK076043.1 | - | 6 | 1147 | |||
AK129469.1 | - | 2488 | ||||
AK167506.1 | - | 6 | 1165 | |||
AK169225.1 | - | 6 | 1154 | |||
BC025907.1 | - | 6 | 1171 | 5064949 |