Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026147.5 | NP_080423.1 | 4 | 370 | ||
GenBank mRNA | AK002405.1 | - | 4 | 371 | ||
AK014594.1 | - | 370 | ||||
AK136398.1 | - | 4 | 373 | |||
AK145656.1 | - | 4 | 371 | |||
AK150867.1 | - | 4 | 370 | |||
AK150972.1 | - | 4 | 370 | |||
BC011323.1 | - | 4 | 357 | 3257215 | ||
BC090389.1 | - | 4 | 361 | 3482252 |