Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000118.3 | NP_000109.1 | 14 | 2873 | ||
NM_001114753.2 | NP_001108225.1 | 15 | 2737 | |||
NM_001278138.1 | NP_001265067.1 | 15 | 2477 | |||
GenBank mRNA | AK222669.1 | - | 12 | 2140 | ||
AK223044.1 | - | 8 | 1300 | |||
AK290389.1 | - | 16 | 2572 | |||
BC014271.2 | - | 15 | 2471 | 3843174 | ||
J05481.1 | - | 15 | 2280 | |||
X72012.1 | - | 14 | 2735 |