Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1lc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026160.4 | NP_080436.1 | 4 | 884 | ||
GenBank mRNA | AF255953.1 | - | 4 | 900 | ||
AK003106.1 | - | 4 | 883 | |||
AK132329.1 | - | 892 | ||||
BC068180.1 | - | 4 | 887 | 30634082 | ||
BC086774.1 | - | 3 | 3311 | |||
BC110308.1 | - | 4 | 873 | 5319360 |