Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286560.1 | NP_001273489.1 | 6 | 746 | ||
NM_026168.4 | NP_080444.1 | 7 | 855 | |||
NM_026355.3 | NP_080631.1 | 7 | 855 | |||
GenBank mRNA | AK004661.1 | - | 7 | 519 | ||
AK007498.1 | - | 7 | 504 | |||
AK008971.1 | - | 7 | 511 | |||
AK009483.1 | - | 7 | 507 | |||
AK010529.1 | - | 7 | 504 | |||
AK014413.1 | - | 7 | 511 | |||
AK016275.1 | - | 7 | 856 | |||
AK017876.1 | - | 7 | 539 | |||
AK030045.1 | - | 7 | 505 | |||
AK086727.1 | - | 7 | 507 | |||
AK135968.1 | - | 4 | 192 | |||
AK162267.1 | - | 6 | 479 | |||
AK167391.1 | - | 7 | 508 | |||
BC006895.1 | - | 7 | 473 | 2651322 | ||
BC018188.1 | - | 7 | 507 | 3711066 | ||
BC026558.1 | - | 7 | 467 | 5030661 | ||
BC064749.1 | - | 7 | 568 | 30046220 |