Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Decr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026172.3 | NP_080448.1 | 2 | 339 | ||
GenBank mRNA | AK002756.1 | - | 2 | 219 | ||
AK004725.2 | - | 2 | 206 | |||
AK010473.1 | - | 2 | 200 | |||
AK015692.1 | - | 2 | 210 | |||
AK146003.1 | - | 2 | 210 | |||
AK168490.1 | - | 2 | 207 | |||
BC046972.1 | - | 2 | 339 | 6469277 | ||
CT010405.1 | - | 2 | 183 |