Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026173.3 | NP_080449.1 | 3, 4 | 477 | ||
GenBank mRNA | AK004769.2 | - | 3, 4 | 405 | ||
AK012926.1 | - | 3, 4 | 445 | |||
AK089013.1 | - | 3, 4 | 413 | |||
AK098149.1 | - | 3, 4 | 370 | |||
AK150656.1 | - | 3, 4 | 478 | |||
BC016527.1 | - | 3, 4 | 391 | 3157281 | ||
BC027118.1 | - | 3, 4 | 415 | 5359153 | ||
BC037631.1 | - | 3, 4 | 434 | 4954391 |