Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253390.1 | NP_001240319.1 | 4 | 352 | ||
NM_001253391.1 | NP_001240320.1 | 4 | 514 | |||
NM_026182.5 | NP_080458.1 | 4 | 362 | |||
GenBank mRNA | AF354708.1 | - | 3 | 313 | ||
AK004846.1 | - | 4 | 340 | |||
AK006952.1 | - | 4 | 514 | |||
AK009195.2 | - | 4 | 328 | |||
AK016316.1 | - | 4 | 500 | |||
AK018048.1 | - | 4 | 344 | |||
AK075822.1 | - | 4 | 338 | |||
AK075992.1 | - | 4 | 362 | |||
AK134822.1 | - | 5 | 356 | |||
AK161251.1 | - | 4 | 335 | |||
BC019971.1 | - | 4 | 310 | 4159093 | ||
CK625955.1 | - | 4 | 342 | |||
DQ369744.1 | - | 4 | 473 |