Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026194.4 | NP_080470.2 | 2 | 239 | ||
GenBank mRNA | AK007993.2 | - | 2 | 184 | ||
AK032663.1 | - | 2 | 175 | |||
AK076206.1 | - | 2 | 184 | |||
AK129211.1 | - | 2 | 181 | |||
AK132697.1 | - | 2 | 203 | |||
BC058809.1 | - | 2 | 178 | |||
BC080203.1 | - | 2 | 217 | |||
BC099529.1 | - | 2 | 227 | |||
BC132195.1 | - | 2 | 191 | 40130472 | ||
BC138153.1 | - | 2 | 191 | 8861172 | ||
BY097965.1 | - | 2 | 241 |