Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290783.1 | NP_001277712.1 | 9 | 1834 | ||
NM_026137.5 | NP_080413.2 | 9 | 1868 | |||
NM_028542.1 | NP_082818.1 | 9 | 2120 | |||
GenBank mRNA | AF220146.1 | - | 8 | 1390 | ||
AF353243.1 | - | 9 | 1514 | |||
AF513713.1 | - | 9 | 1337 | |||
AK006837.1 | - | 8 | 1356 | |||
AK017549.1 | - | 9 | 1450 | |||
AK136979.1 | - | 1 | 34 | |||
BC080853.1 | - | 9 | 1783 | |||
BC108332.1 | - | 9 | 1624 | 6493226 | ||
BC120553.1 | - | 9 | 1371 | 40129476 | ||
BC125287.1 | - | 9 | 1370 | 40129802 |