Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Decr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026172.3 | NP_080448.1 | 10 | 2395 | ||
GenBank mRNA | AK004725.2 | - | 10 | 2261 | ||
AK146003.1 | - | 10 | 2269 | |||
BC046972.1 | - | 10 | 2395 | 6469277 |