Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026175.5 | NP_080451.4 | 15, 16 | 2417 | ||
GenBank mRNA | AK004763.1 | - | 15, 16 | 2400 | ||
AK008075.1 | - | 4, 5 | 444 | |||
AK028829.1 | - | 15, 16 | 2369 | |||
AK029095.1 | - | 15, 16 | 2387 | |||
AK030115.1 | - | 7, 8 | 943 | |||
AK030223.1 | - | 15, 16 | 2404 | |||
AK134178.1 | - | 15, 16 | 2395 | |||
AK160061.1 | - | 15, 16 | 2400 | |||
BC010727.1 | - | 6, 7 | 872 | |||
BC029753.1 | - | 15, 16 | 2381 | 2651773 |