Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026178.2 | NP_080454.1 | 7 | 1472 | ||
GenBank mRNA | AK004819.1 | - | 7 | 1426 | ||
AK011418.1 | - | 7 | 1437 | |||
AK149704.1 | - | 7 | 1425 | |||
AK158577.1 | - | 7 | 1430 | |||
AK171637.1 | - | 7 | 1426 | |||
AY424300.1 | - | 7 | 1312 | |||
BC021914.1 | - | 7 | 1472 | 4506710 | ||
BC034117.1 | - | 8 | 1824 |