Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253390.1 | NP_001240319.1 | 6, 7 | 900 | ||
NM_001253391.1 | NP_001240320.1 | 6, 7 | 1062 | |||
NM_026182.5 | NP_080458.1 | 6, 7 | 910 | |||
GenBank mRNA | AF354708.1 | - | 5, 6 | 861 | ||
AK004846.1 | - | 6, 7 | 888 | |||
AK006952.1 | - | 6, 7 | 1062 | |||
AK009195.2 | - | 6, 7 | 875 | |||
AK012823.1 | - | 888 | ||||
AK016316.1 | - | 6, 7 | 1050 | |||
AK075822.1 | - | 6, 7 | 886 | |||
AK075992.1 | - | 6, 7 | 910 | |||
AK134822.1 | - | 7, 8 | 904 | |||
AK161251.1 | - | 6, 7 | 883 | |||
BC019971.1 | - | 6, 7 | 858 | 4159093 | ||
BC042471.1 | - | 5, 6 | 834 | 3988195 | ||
DQ369744.1 | - | 6, 7 | 1021 |