Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026186.4 | NP_080462.2 | 10 | 2768 | ||
GenBank mRNA | AK004983.2 | - | 10 | 2712 | ||
AK012094.1 | - | 1 | 1161 | |||
AK035158.1 | - | 10 | 2696 | |||
BC053447.1 | - | 3 | 1922 | |||
BC079553.1 | - | 9 | 5440 | 6856335 |