Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026195.3 | NP_080471.2 | 8 | 814 | ||
GenBank mRNA | AK010449.1 | - | 8 | 783 | ||
AK010611.2 | - | 8 | 797 | |||
AK012074.1 | - | 8 | 751 | |||
AK046353.1 | - | 8 | 797 | |||
AK051020.1 | - | 792 | ||||
AK076091.1 | - | 2 | 106 | |||
AK139235.1 | - | 8 | 749 | |||
BC039925.1 | - | 8 | 820 | 3155580 |