Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026200.3 | NP_080476.2 | 3 | 3394 | ||
GenBank mRNA | AK034091.1 | - | 3 | 3287 | ||
AK139482.1 | - | 3 | 3230 |