Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026213.3 | NP_080489.1 | 5 | 1602 | ||
GenBank mRNA | AK010758.1 | - | 5 | 1420 | ||
AK013464.1 | - | 5 | 1448 | |||
AK050426.1 | - | 5 | 1442 | |||
AK075643.1 | - | 5 | 1430 | |||
BC017533.1 | - | 4 | 1367 | |||
BC061018.1 | - | 5 | 1601 | 6590550 |