Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026229.4 | NP_080505.1 | 2, 3 | 176 | ||
GenBank mRNA | AK002937.1 | - | 2, 3 | 196 | ||
AK009734.1 | - | 1, 2 | 55 | |||
AK016787.1 | - | 2, 3 | 176 | |||
BC010729.1 | - | 2, 3 | 169 | 3708143 |
Set Membership: |
G-protein coupled receptor |