Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026240.2 | NP_080516.2 | 4 | 472 | ||
GenBank mRNA | AK018551.1 | - | 5 | 722 | ||
AK018691.1 | - | 4 | 458 | |||
AK038289.1 | - | 3 | 513 | |||
AK161329.1 | - | 4 | 405 | |||
AK164050.1 | - | 4 | 457 | |||
BC057978.1 | - | 4 | 447 | 5044170 |