Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026254.2 | NP_080530.2 | 2 | 193 | ||
GenBank mRNA | AK155142.1 | - | 2 | 214 | ||
AK171591.1 | - | 2 | 101 | |||
BC031706.1 | - | 2 | 193 | 4194266 |