Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026267.2 | NP_080543.2 | 2 | 177 | ||
GenBank mRNA | AK004805.1 | - | 2 | 178 | ||
AK005263.1 | - | 2 | 168 | |||
AK081598.1 | - | 2 | 178 | |||
AK168500.1 | - | 2 | 178 | |||
AK220236.1 | - | 2 | 163 | |||
BC011466.1 | - | 2 | 176 | 4013043 | ||
BC119133.1 | - | 2 | 189 | 8733882 | ||
BC120841.1 | - | 2 | 260 | 40129764 |