Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253920.1 | NP_001240849.1 | 2, 3 | 687 | ||
NM_001290694.1 | NP_001277623.1 | 2, 3 | 769 | |||
NM_026270.4 | NP_080546.1 | 2, 3 | 522 | |||
GenBank mRNA | AK003638.1 | - | 2, 3 | 520 | ||
AK012534.1 | - | 1, 2 | 414 | |||
AK030571.1 | - | 2, 3 | 1065 | |||
AK141771.1 | - | 2, 3 | 518 | |||
AK154110.1 | - | 2, 3 | 504 | |||
BC002180.1 | - | 2, 3 | 689 | |||
BC051563.1 | - | 1, 2 | 410 | |||
BC132372.1 | - | 2, 3 | 471 | 40130649 | ||
BC132374.1 | - | 2, 3 | 471 | 40130651 |