Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026274.4 | NP_080550.3 | 2 | 836 | ||
GenBank mRNA | AK032416.1 | - | 2 | 799 | ||
AK076799.1 | - | 2 | 409 | |||
AK141443.1 | - | 2 | 386 | |||
AK146028.1 | - | 2 | 800 | |||
AK151018.1 | - | 2 | 806 | |||
AK152024.1 | - | 2 | 806 | |||
AK172567.1 | - | 2 | 802 | |||
AK173313.1 | - | 2 | 455 | |||
BC010833.1 | - | 2 | 451 | 4235165 | ||
BC054121.1 | - | 2 | 822 | 6393910 |