Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026275.4 | NP_080551.1 | 1, 2 | 234 | ||
GenBank mRNA | AJ240086.1 | - | 1, 2 | 193 | ||
AK003550.1 | - | 1, 2 | 254 | |||
AK007517.1 | - | 1, 2 | 368 | |||
AK075703.1 | - | 1, 2 | 244 | |||
AK075714.1 | - | 1, 2 | 235 | |||
BC011112.1 | - | 1, 2 | 242 | 3982808 |