Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aasdhppt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026276.3 | NP_080552.3 | 2 | 435 | ||
GenBank mRNA | AK007737.1 | - | 2 | 436 | ||
AK010557.1 | - | 2 | 395 | |||
AK013111.1 | - | 2 | 389 | |||
AK014188.1 | - | 2 | 378 | |||
AK018191.1 | - | 1 | 81 | |||
AK028700.1 | - | 2 | 392 | |||
AK029523.1 | - | 2 | 377 | |||
AK031766.1 | - | 2 | 379 | |||
AK076099.1 | - | 2 | 390 | |||
AK076866.1 | - | 2 | 409 | |||
AK081818.1 | - | 2 | 411 | |||
AK088977.1 | - | 2 | 375 | |||
AK144671.1 | - | 2 | 409 | |||
AK214035.1 | - | 1 | 103 | |||
AK215948.1 | - | 1 | 103 | |||
BC030043.1 | - | 2 | 370 | 1348651 | ||
BC038013.1 | - | 2 | 406 | 3497754 | ||
BC049851.1 | - | 2 | 396 | 3481972 |