Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026323.2 | NP_080599.1 | 3 | 265 | ||
GenBank mRNA | AF334269.1 | - | 3 | 262 | ||
AK005519.1 | - | 3 | 266 | |||
BC099427.1 | - | 3 | 256 | 30845568 |