Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kirrel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190911.1 | NP_001177840.1 | 2 | 489 | ||
NM_001190913.1 | NP_001177842.1 | 2 | 489 | |||
NM_001190914.1 | NP_001177843.1 | 3 | 590 | |||
NM_026324.3 | NP_080600.1 | 2 | 489 | |||
GenBank mRNA | AK034142.1 | - | 2 | 426 | ||
AK038385.1 | - | 2 | 240 | |||
AK045373.1 | - | 2 | 394 | |||
AK079332.1 | - | 3 | 527 | |||
AY169782.1 | - | 2 | 489 |