Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026330.3 | NP_080606.2 | 3 | 311 | ||
GenBank mRNA | AK009715.1 | - | 3 | 200 | ||
AK011011.1 | - | 3 | 216 | |||
AK160217.1 | - | 3 | 211 | |||
AK166410.1 | - | 3 | 204 | |||
BC031848.1 | - | 3 | 290 | 3984224 | ||
BC049558.1 | - | 3 | 304 | 6707952 |