Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164237.1 | NP_001157709.1 | 6 | 946 | ||
NM_026259.3 | NP_080535.2 | 6 | 946 | |||
GenBank mRNA | AK015745.1 | - | 5 | 666 | ||
AK016825.1 | - | 5 | 526 | |||
AK020527.1 | - | 4 | 519 | |||
AK028551.1 | - | 6 | 817 | |||
AK030558.1 | - | 6 | 945 | |||
AK031657.1 | - | 5 | 588 | |||
AK034551.1 | - | 6 | 862 | |||
AK044103.1 | - | 5 | 578 | |||
AK045760.1 | - | 6 | 930 | |||
AK077466.1 | - | 6 | 805 | |||
BC019415.1 | - | 3 | 315 | 4014525 | ||
BC049078.1 | - | 6 | 905 | 6419123 |