Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026267.2 | NP_080543.2 | 5 | 416 | ||
GenBank mRNA | AK004805.1 | - | 5 | 417 | ||
AK005263.1 | - | 5 | 407 | |||
AK081598.1 | - | 5 | 417 | |||
AK168500.1 | - | 5 | 417 | |||
AK211496.1 | - | 2 | 105 | |||
AK220236.1 | - | 5 | 402 | |||
BC011466.1 | - | 5 | 415 | 4013043 | ||
BC119133.1 | - | 5 | 428 | 8733882 | ||
BC120841.1 | - | 5 | 499 | 40129764 |