Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: l7Rn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291286.1 | NP_001278215.1 | 3 | 488 | ||
NM_001291287.1 | NP_001278216.1 | 4 | 552 | |||
NM_001291288.1 | NP_001278217.1 | 4 | 629 | |||
NM_001291289.1 | NP_001278218.1 | 2 | 250 | |||
NM_026304.3 | NP_080580.1 | 3 | 488 | |||
GenBank mRNA | AK002306.1 | - | 3 | 470 | ||
AK003324.1 | - | 2 | 241 | |||
AK008604.1 | - | 2 | 252 | |||
AK012166.1 | - | 3 | 454 | |||
AK206361.1 | - | 2 | 263 | |||
BC003916.1 | - | 3 | 436 | 3485003 | ||
BC083330.1 | - | 4 | 490 | |||
DQ196315.1 | - | 3 | 466 | |||
DQ196316.1 | - | 3 | 466 |